missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CXCL2/GRO beta/MIP-2/CINC-3 aa 39-107 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 2920 |
Molecular Weight (g/mol) | 7.5 kDa |
Name | CXCL2/GRO beta/MIP-2/CINC-3 aa 39-107 |
Quantity | 25μg |
Description
The Recombinant Human CXCL2/GRO beta (aa 39-107) Protein is derived from E. coli. The Recombinant Human CXCL2/GRO beta (aa 39-107) Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
7.5 kDa | |
25μg | |
E. coli-derived human CXCL2/GRO beta/MIP-2/CINC-3 protein Thr39-Asn107 |
2920 | |
CXCL2/GRO beta/MIP-2/CINC-3 aa 39-107 | |
Yes |