missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CXCL16 Chemokine Domain Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 58191 |
Molecular Weight (g/mol) | 10.2 kDa |
Name | CXCL16 Chemokine Domain |
Quantity | 25μg |
Description
The Recombinant Human CXCL16 Protein is derived from E. coli. The Recombinant Human CXCL16 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
10.2 kDa | |
25μg | |
E. coli-derived human CXCL16 protein Asn49-Pro137 |
58191 | |
CXCL16 Chemokine Domain | |
Yes |