missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL10/IP-10/CRG-2 Recombinant Protein

Brand:  R&D Systems™ 266-IP-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461694

  • £391.31 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Val22-Pro98, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3627
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3627
CXCL10/IP-10/CRG-2
50μg
Unlabeled
Videos