missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CTLA-4 Fc Chimera Recombinant Protein

Brand:  R&D Systems™ 325-CT-200/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462674

  • £391.31 / 200µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala37-Phe162, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 40 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1493
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
40 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
1493
CTLA-4 Fc Chimera
200μg
Disulfide-linked homodimer
Yes
Videos