missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Crossveinless-2/CV-2 Recombinant Protein

Brand:  R&D Systems™ 1956-CV-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460294

  • £316.81 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Val34-Asp369, with an N-terminal 7-His tag & Ala39-Asp369 (N-terminal fragments) & Pro370-Arg685 (C-terminal fragment)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 168667
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
Crossveinless-2/CV-2
50μg
Disulfide-linked heterodimer
Yes
168667
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
Videos