missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Crossveinless-2/CV-2 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 168667 |
Name | Crossveinless-2/CV-2 |
Quantity | 50μg |
Recombinant | Yes |
Description
The Recombinant Human Crossveinless-2 Protein is derived from NS0. The Recombinant Human Crossveinless-2 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
Crossveinless-2/CV-2 | |
Yes |
168667 | |
50μg | |
Mouse myeloma cell line,NS0-derived human Crossveinless-2/CV-2 protein Val34-Asp369,with an N-terminal 7-His tag & Ala39-Asp369 (N-terminal fragments) & Pro370-Arg685 (C-terminal fragment) |