missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CLF-1/CLC Complex Recombinant Protein

Brand:  R&D Systems™ 1151-CL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458743

  • £298.33 / 25µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala38-Arg422, with a C-terminal 10-His tag (CLF-1); Leu28-Phe225, with a C-terminal 6-His tag (CLC)
Purity: 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 44.5 kDa (CLF-1) & 23 kDa (CLC)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 9244
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
44.5 kDa (CLF-1) & 23 kDa (CLC)
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
9244
CLF-1/CLC Complex
25μg
Unlabeled
Videos