missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CLF-1/CLC Complex Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1151-CL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468743

  • £342.51 / 25µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human CLF-1/CLC Complex Protein is derived from NS0. The Recombinant Human CLF-1/CLC Complex Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
44.5 kDa (CLF-1) & 23 kDa (CLC)
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
9244
CLF-1/CLC Complex
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents