missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CLC Recombinant Protein

Brand:  R&D Systems™ 962-CL-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486454

  • £356.00 / 50µg


Estimated Shipment
28-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Leu28-Phe225, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 23529
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
22.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
23529
CLC
50μg
Unlabeled
Videos