missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CLC Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Gene ID (Entrez) | 23529 |
---|---|
Molecular Weight (g/mol) | 22.4 kDa |
Name | CLC |
Quantity | 50μg |
Source | E. coli-derived human CLC protein Leu28-Phe225,with an N-terminal Met |
Description
The Recombinant Human CLC Protein is derived from E. coli. The Recombinant Human CLC Protein has been validated for the following applications: Bioactivity.Specifications
23529 | |
CLC | |
E. coli-derived human CLC protein Leu28-Phe225,with an N-terminal Met | |
Unconjugated | |
CLC |
22.4 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |