missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD8 alpha+/IFN-gamma ELISpot Kit

For the quantitative determination of the frequency of CD8 alpha+ cells releasing human IFN-gamma

Brand:  R&D Systems™ EL3094

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465105

  • £443.94 / 1 set


Estimated Shipment date
from Supplier 02-04-2020
Add to basket

Description

Description

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • Hig
Specifications

Specifications

Diagnostics
Human CD8 alpha+/IFN-gamma ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
One 96-well PVDF-backed plate coated w/anti-human CD8a+ and anti-human IFNG mAb, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive Ctrl
Whole Cells
One 96-well plate
Videos
Documents