missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CD8 alpha+/IFN-gamma ELISpot Kit
For the quantitative determination of the frequency of CD8 alpha+ cells releasing human IFN-gamma
POA
Specifications
Product Type | Human CD8 alpha+/IFN-gamma ELISpot Kit |
---|---|
For Use With (Application) | Diagnostics |
Sample Type | Whole Cells |
Description
Product Features:- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- Hig
Specifications
Human CD8 alpha+/IFN-gamma ELISpot Kit | |
Diagnostics | |
Whole Cells |