missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD55/DAF Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 2009-CD-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440394

  • £290.08 / 50µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human CD55/DAF Protein is derived from NS0. The Recombinant Human CD55/DAF Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
36.0 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line,NS0-derived human CD55/DAF protein, (N-terminus) Human CD55 (Asp35-Ser353) Accession &Num; P08174.4 DI 6-His tag (C-terminus)
Unconjugated
1604
CD55/DAF
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents