missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD40 Ligand/TNFSF5 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 6420-CL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13455794

  • £366.03 / 25µg

Estimated Shipment date
from Supplier 05-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human CD40 Ligand/TNFSF5 (HEK293-expressed) is derived from HEK293. The Recombinant Human CD40 Ligand/TNFSF5 (HEK293-expressed) has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
21.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human embryonic kidney cell, HEK293-derived Hemagglutinin tag YPYDVPDYA GCN4-IZ (Met1-Ile29) GGGSGGGSGGGS Human CD40 Ligand (Met113-Leu261) Accession # P29965
Unlabeled
959
CD40 Ligand/TNFSF5
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents