missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD40 Ligand/TNFSF5 Recombinant Protein

Brand:  R&D Systems™ 6420-CL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465794

  • £319.45 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Human embryonic kidney cell, HEK293-derived, Met113-Leu261
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and EDTA.
Entrez Gene ID: 959
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and EDTA.
21.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human embryonic kidney cell, HEK293-derived Hemagglutinin tag YPYDVPDYA GCN4-IZ (Met1-Ile29) GGGSGGGSGGGS Human CD40 Ligand (Met113-Leu261) Accession # P29965
Yes
959
CD40 Ligand/TNFSF5
25μg
Unlabeled
Videos