missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD40 Ligand/TNFSF5 (Histidine-tagged) Recombinant Protein

Brand:  R&D Systems™ 2706-CL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461764

  • £182.16 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Glu108-Leu261, with an N-terminal Met and 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Phosphate, NaCl and EDTA.
Entrez Gene ID: 959
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and EDTA.
17.7 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
959
CD40 Ligand/TNFSF5 (Histidine-tagged)
25μg
Homotrimer
Yes
Videos