missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD40 Ligand/TNFSF5 aa 108-261 Recombinant Protein

Brand:  R&D Systems™ 6245-CL-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465744

  • £335.29 / 50µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: E. coli-derived, Glu108-Leu261, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.9 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA.
Entrez Gene ID: 959
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA.
16.9 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
959
CD40 Ligand/TNFSF5 aa 108-261
50μg
Unlabeled
Videos