missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human sCD4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 920 |
---|---|
Molecular Weight (g/mol) | 40 kDa |
Name | CD4 |
Quantity | 50μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human CD4 protein Lys26-Trp390 |
Description
Measured by the ability of the immobilized protein to support the adhesion of HeLa human cervical epithelial carcinoma cells. The ED50 for this effect is 1-8 μg/mL. Optimal dilutions should be determined by each laboratory for each application.Specifications
920 | |
CD4 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human CD4 protein Lys26-Trp390 | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
40 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |