missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD25/IL-2 R alpha Recombinant Protein

Brand:  R&D Systems™ 223-2A-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460844

  • £578.52 / 25µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Glu22-Cys213
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3559
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
22 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3559
CD25/IL-2 R alpha
25μg
Unlabeled
Videos