missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL5/RANTES Recombinant Protein, N-terminal Met

Brand:  R&D Systems™ 335-RM-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462884

  • £220.44 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ser24-Ser91, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6352
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
7.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6352
CCL5/RANTES
25μg
Unlabeled
Videos