missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL3/MIP-1 alpha Isoform LD78a Recombinant Protein

Brand:  R&D Systems™ 270-LD-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461774

  • £190.08 / 10µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala27-Ala92
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6348
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6348
CCL3/MIP-1 alpha Isoform LD78a
10μg
Unlabeled
Videos