missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL3/MIP-1 alpha Isoform LD78a Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 6348 |
Molecular Weight (g/mol) | 7.5 kDa |
Name | CCL3/MIP-1 alpha Isoform LD78a |
Recombinant | Yes |
Description
The Recombinant Human CCL3/MIP-1 alpha Protein is derived from E. coli. The Recombinant Human CCL3/MIP-1 alpha Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
7.5 kDa | |
Yes |
6348 | |
CCL3/MIP-1 alpha Isoform LD78a | |
E. coli-derived human CCL3/MIP-1 alpha protein Ala27-Ala92 |