missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL24/Eotaxin-2/MPIF-2 Recombinant Protein

Brand:  R&D Systems™ 343-E2-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463024

  • £182.16 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Val27-Cys119
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6369
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
10.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6369
CCL24/Eotaxin-2/MPIF-2
25μg
Unlabeled
Videos