missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CCL23/MPIF-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 6368 |
---|---|
Molecular Weight (g/mol) | 11.5 kDa |
Name | CCL23/MPIF-1 |
Quantity | 25μg |
Source | E. coli-derived human CCL23/MPIF-1 protein Arg22-Asn120 |
Description
The Recombinant Human CCL23/MPIF-1 Protein is derived from E. coli. The Recombinant Human CCL23/MPIF-1 Protein has been validated for the following applications: Bioactivity.Specifications
6368 | |
CCL23/MPIF-1 | |
E. coli-derived human CCL23/MPIF-1 protein Arg22-Asn120 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
11.5 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |