missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL22/MDC Recombinant Protein

Brand:  R&D Systems™ 336-MD-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462904

  • £355.09 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Gly25-Gln93
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6367
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6367
CCL22/MDC
25μg
Unlabeled
Videos