missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL21/6Ckine Recombinant Protein

Brand:  R&D Systems™ 366-6C-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463384

  • £343.21 / 25µg


Estimated Shipment
22-10-2018
Add to basket

Description

Description

Source: E. coli-derived, Ser24-Pro134
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6366
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
12 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
6366
CCL21/6Ckine
25μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos