missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL18/PARC Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 394-PA-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13433874

  • £249.90 / 10µg

Estimated Shipment date
from Supplier 27-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human CCL18/PARC Protein is derived from E. coli. The Recombinant Human CCL18/PARC Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
6362
CCL18/PARC
10μg
12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution.
Yes
Videos
Documents