missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL18/PARC Recombinant Protein

Brand:  R&D Systems™ 394-PA-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463874

  • £316.81 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala21-Ala89
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6362
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 100μg/mL in sterile PBS.
6362
CCL18/PARC
50μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in Acetonitrile with Trehalose.
7.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
Videos