missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL17/TARC Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

POA

Specifications

Conjugate Unconjugated
Gene ID (Entrez) 6361
Molecular Weight 8 kDa
Name CCL17/TARC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs

Description

Description

The Recombinant Human CCL17/TARC Protein is derived from E. coli.
Specifications

Specifications

Unconjugated
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6361
CCL17/TARC
25μg
E. coli-derived human CCL17/TARC protein Ala24-Ser94