missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL17/TARC Recombinant Protein

Brand:  R&D Systems™ 364-DN-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463354

  • £341.89 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala24-Ser94
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6361
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6361
CCL17/TARC
25μg
Unlabeled
Videos