missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Caspase-3 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 707-C3-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466054

  • £308.70 / 10µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Caspase-3 Protein is derived from E. coli. The Recombinant Human Caspase-3 Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose with BSA as a carrier protein.
17 kDa (subunit 1), 11 kDa (subunit 2)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
836
Caspase-3
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes
Videos
Documents