missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human C-Reactive Protein/CRP Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 1707-CR-200

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468673

  • £289.59 / 200µg

Estimated Shipment date
from Supplier 04-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human C-Reactive Protein/CRP Protein is derived from NS0. The Recombinant Human C-Reactive Protein/CRP Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 with BSA as a carrier protein.
23 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Homopentamer
Yes
1401
C-Reactive Protein/CRP
200μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents