missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-8a Recombinant Protein

Brand:  R&D Systems™ 1073-BP-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458453

  • £320.77 / 10µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala264-His402 & Val265-His402
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 656
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
15.8 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
656
BMP-8a
10μg
Disulfide-linked homodimer
Yes
Videos