missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-5 Recombinant Protein, (CHO-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 615-BMC-020

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445704

  • £361.62 / 20µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human BMP-5 (CHO-expressed) Protein is derived from CHO. The Recombinant Human BMP-5 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
15.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
653
BMP-5
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents