missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-4 Recombinant Protein

Brand:  R&D Systems™ 314-BP-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462534

  • £1030.35 / 50µg


Estimated Shipment
01-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ser293-Arg408
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 13 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 652
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
13 kDa (monomer)
95%, by SDS-PAGE with silver staining.
Human
Unlabeled
652
BMP-4
50μg
Disulfide-linked homodimer
Yes
Videos