missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-4 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 314-BP-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13452534

  • £457.17 / 10µg

Estimated Shipment date
from Supplier 02-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human BMP-4 Protein is derived from NS0. The Recombinant Human BMP-4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
13 kDa (monomer)
95%, by SDS-PAGE with silver staining.
Human
Disulfide-linked homodimer
Unlabeled
652
BMP-4
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -70°C under sterile conditions after reconstitution.
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Yes
Videos
Documents