missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-3b/GDF-10 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1543-BP-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439643

  • £518.00 / 25µg

Estimated Shipment
11-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human BMP-3b/GDF-10 Protein is derived from E. coli. The Recombinant Human BMP-3b/GDF-10 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
12.5 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human BMP-3b/GDF-10 protein Gln369-Arg478,with an N-terminal Met
Disulfide-linked homodimer
Yes
2662
BMP-3b/GDF-10
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Videos
Documents