missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-2 Recombinant Protein, (E. coli-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 355-BEC-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463194

  • £220.52 / 10µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed) is derived from E. coli. The Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed) has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
12.8 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived BMP-2 protein Ala284-Arg396 This form is not detected using human BMP-2 Quantikine ELISA (Catalog &Num; DBP200 ).
Disulfide-linked homodimer
Unconjugated
650
BMP-2
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Yes
Videos
Documents