missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-2/BMP-7 Heterodimer Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 3229-BM-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13452604

  • £514.50 / 10µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human BMP-2/BMP-7 Heterodimer Protein is derived from E. coli. The Recombinant Human BMP-2/BMP-7 Heterodimer Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
12.9 kDa (BMP-2), 15.8 kDa (BMP-7)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer
Yes
650
BMP-2/BMP-7 Heterodimer
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents