missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-2/BMP-7 Heterodimer Recombinant Protein

Brand:  R&D Systems™ 3229-BM-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462604

  • £442.02 / 10µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala284-Arg396 (BMP-2) & Ser293-His431 (BMP-7)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.9 kDa (BMP-2), 15.8 kDa (BMP-7)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 650
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
12.9 kDa (BMP-2), 15.8 kDa (BMP-7)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
650
BMP-2/BMP-7 Heterodimer
10μg
Disulfide-linked heterodimer
Yes
Videos