missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Betacellulin/BTC Recombinant Protein

Brand:  R&D Systems™ 261-CE-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461584

  • £918.99 / 50µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Asp32-Tyr111
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 685
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
685
Betacellulin/BTC
50μg
Unlabeled
Videos