missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BAFF/BLyS/TNFSF13B Recombinant Protein

Brand:  R&D Systems™ 2149-BF-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460664

  • £174.24 / 10µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala134-Leu285
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Entrez Gene ID: 10673
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
19.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived MHHHHHHHHHH GGGSGGGSGGGS IEGR Human BAFF (Ala134-Leu285) Accession # Q9Y275
Yes
10673
BAFF/BLyS/TNFSF13B
10μg
Unlabeled
Videos