missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human APRIL/TNFSF13 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 5860-AP-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445584

  • £282.24 / 10µg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human APRIL/TNFSF13 (HEK293-expressed) Protein is derived from HEK293. The Recombinant Human APRIL/TNFSF13 (HEK293-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Bioactivity
8741
APRIL/TNFSF13
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protin.
21.7 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Noncovalently-linked homotrimer
Yes
Videos
Documents