missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Angiopoietin-like Protein 7/ANGPTL7 Recombinant Protein

Brand:  R&D Systems™ 914-AN-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486404

  • £338.00 / 25µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-Pro346, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 38.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 10218
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
38.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
10218
Angiopoietin-like Protein 7/ANGPTL7
25μg
Unlabeled
Videos