missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Angiopoietin-1 Recombinant Protein

Brand:  R&D Systems™ 923-AN-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486414

  • £317.00 / 25µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ser20-Phe498, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 56 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
Entrez Gene ID: 284
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl with BSA as a carrier protein.
56 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
284
Angiopoietin-1
25μg
Oligomer
Yes
Videos