missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Amphiregulin Recombinant Protein

Brand:  R&D Systems™ 262-AR-100/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461604

  • £340.57 / 100µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ser101-Lys198
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 374
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
11 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
374
Amphiregulin
100μg
Unlabeled
Videos