missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Activin AC Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 4879-AC-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13435144

  • £342.51 / 10µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Activin AC Heterodimer Protein is derived from CHO. The Recombinant Human Activin AC Heterodimer Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Activin AC
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer
Yes
Videos
Documents