missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Activin AB Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1066-AB-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428433

  • £363.09 / 5µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Activin AB Protein is derived from CHO. The Recombinant Human Activin AB Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Activin AB
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.1 EU per 1μg of the protein by the LAL method.
Yes
12.8 kDa (betaA subunit) and 13 kDa (betaB subunit)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer
Unlabeled
Videos
Documents