missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Activin AB Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Molecular Weight (g/mol) | 12.8 kDa (betaA subunit) and 13 kDa (betaB subunit) |
Name | Activin AB |
Quantity | 5μg |
Recombinant | Yes |
Description
The Recombinant Human Activin AB Protein is derived from CHO. The Recombinant Human Activin AB Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
Activin AB | |
Yes |
12.8 kDa (betaA subunit) and 13 kDa (betaB subunit) | |
5μg | |
Chinese Hamster Ovary cell line,CHO-derived human Activin AB protein, (N-terminus) Human Activin beta A (Gly311-Ser426) Accession &Num; NP_002183.1 Human Activin beta B (Gly293-Ala407) Accession &Num; NP_002184.2 (C-terminus) |