Learn More
R&D Systems™ Feline IL-5 Recombinant Protein
Brand: R&D Systems™ 1795-FL-010
Code : Z1
Additional Details : Weight : 0.00010kg
Description
Source: Mouse myeloma cell line, NS0-derived, Ile20-Ser134, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493803
Regulatory Status: RUO
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
14.6 kDa | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. | |
Feline | |
Unlabeled |
493803 | |
IL-5 | |
10μg | |
Disulfide-linked homodimer | |
Yes |